Assay Details
Target Gene Details
Entrez Gene ID: | 146712 |
Gene Name: | UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase-like 1 |
Gene Aliases: |
3-Gn-T8, B3GNT8, BGnT-8, beta-1, beta3Gn-T8, beta3GnTL1 |
Location: |
Chr.17:82942149-83051845 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
B3GNTL1 | NM_001009905.2 | NP_001009905.1 | ||
NM_001320742.1 | NP_001307671.1 | |||
NM_001320743.1 | NP_001307672.1 | |||
NR_135465.1 | ||||
NR_135466.1 | ||||
XM_006722272.3 | XP_006722335.1 | |||
XM_011523532.2 | XP_011521834.1 | |||
XM_011523535.1 | XP_011521837.1 | |||
XM_017024205.1 | XP_016879694.1 | |||
XM_017024206.1 | XP_016879695.1 | |||
AK054757.1 | ||||
AK126018.1 | ||||
AY304503.1 | AAQ74775.1 | |||
AY634364.1 | AAT47555.1 | |||
BC019623.1 | AAH19623.1 | |||
U79265.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1067553 | Chr.17:82530808 - 83091923 on Build GRCh38 | Loss | LOC105371944 FN3K TBCD FN3KRP WDR45B MIR4525 RAB40B ZNF750 LOC105376791 METRNL FOXK2 LOC101929552 B3GNTL1 |
dgv3296n100 | Chr.17:82851564 - 83091923 on Build GRCh38 | Gain | LOC105371944 METRNL TBCD B3GNTL1 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | SIRT7 CEP131 P4HB FASN OGFOD3 ARL16 SLC25A10 FN3KRP AATK ACTG1 TSPAN10 AATK-AS1 ZNF750 SLC16A3 ASPSCR1 LOC105376791 CCDC57 MIR6786 C17orf89 MYADML2 B3GNTL1 CSNK1D TEPSIN TEX19 NPLOC4 GPS1 SLC38A10 ALYREF NOTUM WDR45B HGS MIR4525 RAB40B MIR3186 FAAP100 NPB MAFG FOXK2 ARHGDIA LOC100130370 LOC105371944 FN3K PPP1R27 DUS1L LOC105371925 MIR6787 OXLD1 LOC101929511 RAC3 TBCD MIR4740 HEXDC LRRC45 CD7 MCRIP1 RFNG PYCR1 PDE6G NARF PCYT2 C17orf62 LOC105376790 MRPL12 UTS2R BAHCC1 MAFG-AS1 GCGR ANAPC11 DCXR LINC00482 FSCN2 STRA13 TMEM105 LOC101929552 SECTM1 SNORD134 CCDC137 |
nsv576217 | Chr.17:82937869 - 82960820 on Build GRCh38 | Loss | LOC105371944 TBCD B3GNTL1 |
dgv3297n100 | Chr.17:82879855 - 83048753 on Build GRCh38 | Gain | LOC105371944 TBCD B3GNTL1 |
esv3893052 | Chr.17:82895301 - 83102003 on Build GRCh38 | Gain | LOC105371944 METRNL TBCD B3GNTL1 |
nsv1057153 | Chr.17:82876571 - 83091923 on Build GRCh38 | Loss | LOC105371944 METRNL TBCD B3GNTL1 |
esv21913 | Chr.17:81828123 - 83103548 on Build GRCh38 | Gain+Loss | SIRT7 P4HB FASN OGFOD3 FN3KRP ZNF750 SLC16A3 ASPSCR1 LOC105376791 CCDC57 MYADML2 B3GNTL1 CSNK1D TEX19 GPS1 ALYREF NOTUM WDR45B MIR4525 RAB40B NPB MAFG FOXK2 ARHGDIA LOC105371944 FN3K PPP1R27 DUS1L MIR6787 LOC101929511 RAC3 TBCD HEXDC LRRC45 CD7 MCRIP1 METRNL RFNG PYCR1 NARF PCYT2 C17orf62 LOC105376790 UTS2R MAFG-AS1 ANAPC11 DCXR STRA13 LOC101929552 SECTM1 SNORD134 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |