Assay Details
Target Gene Details
Entrez Gene ID: | 23210 |
Gene Name: | arginine demethylase and lysine hydroxylase |
Gene Aliases: |
PSR, PTDSR, PTDSR1 |
Location: |
Chr.17:76712830-76726799 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
JMJD6 | NM_001081461.1 | NP_001074930.1 | ||
NM_015167.2 | NP_055982.2 | |||
AB011157.1 | ||||
AB073711.1 | BAC16755.1 | |||
AK021780.1 | ||||
AK294816.1 | ||||
AK314541.1 | ||||
BC047003.1 | AAH47003.1 | |||
BC066654.1 | AAH66654.1 | |||
EF527406.1 | ||||
EF527407.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833550 | Chr.17:76712038 - 76886290 on Build GRCh38 | Loss | METTL23 SRSF2 JMJD6 MIR636 MGAT5B MFSD11 LINC00868 LOC101928514 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |