Assay Details
Target Gene Details
Entrez Gene ID: | 8288 |
Gene Name: | eosinophil peroxidase |
Gene Aliases: |
EPO, EPP, EPX-PEN, EPXD |
Location: |
Chr.17:58192724-58205174 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPX | NM_000502.5 | NP_000493.1 | ||
X14346.1 | CAA32530.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517229 | Chr.17:58176226 - 58373163 on Build GRCh38 | Gain+Loss | MIR142 TSPOAP1 RNF43 EPX SUPT4H1 MPO MKS1 TSPOAP1-AS1 LPO MIR4736 |
nsv457858 | Chr.17:58180623 - 58212973 on Build GRCh38 | Loss | EPX MKS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |