Assay Details
Target Gene Details
Entrez Gene ID: | 64750 |
Gene Name: | SMAD specific E3 ubiquitin protein ligase 2 |
Gene Aliases: |
- |
Location: |
Chr.17:64542282-64662268 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMURF2 | NM_022739.3 | NP_073576.1 | ||
XM_005257585.3 | XP_005257642.1 | |||
XM_011525128.1 | XP_011523430.1 | |||
AF301463.1 | AAG25641.1 | |||
AF310676.1 | AAG45422.1 | |||
AY014180.1 | AAG50421.1 | |||
BC009527.1 | AAH09527.1 | |||
BC093876.1 | AAH93876.1 | |||
BC111945.1 | AAI11946.1 | |||
CR936721.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1066765 | Chr.17:64520760 - 65128185 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)