Assay Details
Target Gene Details
Entrez Gene ID: | 105369205 |
Gene Name: | uncharacterized LOC105369205 |
Gene Aliases: |
- |
Location: |
Chr.17:20921016-21001059 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105369205 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1064365 | Chr.17:20898426 - 20987403 on Build GRCh38 | Gain | LOC339260 CCDC144NL-AS1 LOC105369205 |
dgv5502n54 | Chr.17:20899465 - 20969018 on Build GRCh38 | Gain | LOC339260 CCDC144NL-AS1 LOC105369205 |
esv2751619 | Chr.17:20559595 - 21010843 on Build GRCh38 | Gain | LOC339260 MEIS3P2 ABHD17AP6 CCDC144NL-AS1 CDRT15L2 LOC105369205 USP22 LOC100287072 CCDC144NL |
nsv1059146 | Chr.17:20849194 - 22546654 on Build GRCh38 | Gain | LOC100996792 CCDC144NL-AS1 LINC01563 KCNJ18 NATD1 C17orf51 LOC339260 TMEM11 UBBP4 DHRS7B MTRNR2L1 LOC105369205 MAP2K3 USP22 CCDC144NL FLJ36000 FAM27E5 KCNJ12 |
esv3640221 | Chr.17:20913840 - 20955637 on Build GRCh38 | Loss | LOC339260 LOC105369205 |
nsv457706 | Chr.17:20911453 - 20969018 on Build GRCh38 | Gain | LOC339260 LOC105369205 |