Assay Details
Target Gene Details
Entrez Gene ID: | 388336 |
Gene Name: | shisa family member 6 |
Gene Aliases: |
- |
Location: |
Chr.17:11241423-11564063 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SHISA6 | NM_001173461.1 | NP_001166932.1 | ||
NM_001173462.1 | NP_001166933.1 | |||
NM_207386.3 | NP_997269.2 | |||
XM_017024618.1 | XP_016880107.1 | |||
AK127379.1 | BAC86949.1 | |||
DA362563.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv574359 | Chr.17:11223935 - 11275051 on Build GRCh38 | Loss |
![]() |
nsv1057473 | Chr.17:11182844 - 11287592 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)