Assay Details
Target Gene Details
Entrez Gene ID: | 84690 |
Gene Name: | spermatogenesis associated 22 |
Gene Aliases: |
NYD-SP20, NYDSP20 |
Location: |
Chr.17:3440011-3513852 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPATA22 | NM_001170695.1 | NP_001164166.1 | ||
NM_001170696.1 | NP_001164167.1 | |||
NM_001170697.1 | NP_001164168.1 | |||
NM_001170698.1 | NP_001164169.1 | |||
NM_001170699.1 | NP_001164170.1 | |||
NM_001321336.1 | NP_001308265.1 | |||
NM_001321337.1 | NP_001308266.1 | |||
NM_032598.4 | NP_115987.2 | |||
AF367472.1 | AAK53408.1 | |||
AK295299.1 | ||||
AK301892.1 | ||||
AY032684.1 | AAK51120.1 | |||
AY035867.1 | AAK61373.1 | |||
AY035868.1 | AAK61374.1 | |||
BC029483.1 | AAH29483.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892950 | Chr.17:3131812 - 3577153 on Build GRCh38 | Loss | OR1A1 OR1A2 OR1D4 SPATA22 TRPV3 OR3A4P OR3A3 OR3A1 OR1E1 TRPV1 ASPA OR1E2 OR3A2 |
nsv1065489 | Chr.17:2631416 - 3948349 on Build GRCh38 | Gain | OR1A2 MIR1253 EMC6 OR1D4 SPATA22 P2RX5 OR1D2 OR3A3 LOC101927911 TAX1BP3 OR1G1 CLUH TRPV1 ASPA CCDC92B LOC105371591 P2RX5-TAX1BP3 GSG2 RAP1GAP2 MIR6776 OR1A1 ATP2A3 NCBP3 TRPV3 OR3A4P OR1D5 PAFAH1B1 ITGAE LOC105371592 OR3A1 SHPK OR1E1 OR1E2 P2RX1 CTNS CAMKK1 OR3A2 |
nsv574232 | Chr.17:3254715 - 3503170 on Build GRCh38 | Gain | OR3A1 OR1E1 ASPA SPATA22 OR1E2 OR3A4P OR3A2 OR3A3 |
esv2660206 | Chr.17:2679367 - 3519614 on Build GRCh38 | Deletion | RAP1GAP2 MIR6776 OR1A1 OR1A2 MIR1253 OR1D4 SPATA22 TRPV3 OR3A4P OR1D2 OR3A3 OR1D5 LOC101927911 PAFAH1B1 LOC105371592 OR3A1 OR1E1 OR1G1 CLUH ASPA OR1E2 CCDC92B LOC105371591 OR3A2 |
nsv1055740 | Chr.17:3381841 - 3890153 on Build GRCh38 | Gain | GSG2 EMC6 SPATA22 NCBP3 P2RX5 TRPV3 OR3A3 TAX1BP3 ITGAE SHPK OR1E1 TRPV1 ASPA OR1E2 CTNS CAMKK1 OR3A2 P2RX5-TAX1BP3 |
nsv521277 | Chr.17:3390044 - 3468885 on Build GRCh38 | Gain | OR1E1 SPATA22 OR1E2 OR3A3 |
nsv833341 | Chr.17:3352243 - 3521390 on Build GRCh38 | Loss | OR1E1 ASPA SPATA22 OR1E2 TRPV3 OR3A2 OR3A3 |
nsv1060071 | Chr.17:3398474 - 3633472 on Build GRCh38 | Loss | SHPK TRPV1 ASPA SPATA22 OR1E2 TRPV3 OR3A3 |
nsv1059413 | Chr.17:3365789 - 3453922 on Build GRCh38 | Loss | OR1E1 SPATA22 OR1E2 OR3A2 OR3A3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |