Assay Details
Target Gene Details
Entrez Gene ID: | 6352 |
Gene Name: | C-C motif chemokine ligand 5 |
Gene Aliases: |
D17S136E, RANTES, SCYA5, SIS-delta, SISd, TCP228, eoCP |
Location: |
Chr.17:35871491-35880373 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCL5 | NM_001278736.1 | NP_001265665.1 | ||
NM_002985.2 | NP_002976.2 | |||
AF043341.1 | AAC03541.1 | |||
AF266753.1 | AAF73070.1 | |||
AK312212.1 | ||||
BC008600.1 | AAH08600.1 | |||
CR980938.1 | ||||
DQ230537.1 | ||||
M21121.1 | AAA36725.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059795 | Chr.17:35828724 - 36157706 on Build GRCh38 | Gain | HEATR9 CCL16 CCL4 LYZL6 CCL5 CCL15-CCL14 CCL23 CCL14 LRRC37A8P LOC101927339 CCL18 RDM1 CCL15 CCL3 TAF15 |
esv3892999 | Chr.17:35252863 - 35925533 on Build GRCh38 | Loss | SNORD7 HEATR9 C17orf50 LOC105371743 PEX12 SLFN12 CCL5 RASL10B LOC105371933 SLFN12L SLFN11 SLFN14 MMP28 SLFN13 LRRC37A8P AP2B1 RDM1 GAS2L2 SLFN5 TAF15 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |