Assay Details
Target Gene Details
Entrez Gene ID: | 3675 |
Gene Name: | integrin subunit alpha 3 |
Gene Aliases: |
CD49C, FRP-2, GAP-B3, GAPB3, ILNEB, MSK18, VCA-2, VL3A, VLA3a |
Location: |
Chr.17:50055968-50090485 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ITGA3 | NM_002204.3 | NP_002195.1 | ||
XM_005257308.2 | XP_005257365.1 | |||
AB209658.1 | BAD92895.1 | |||
AK289961.1 | ||||
AK293323.1 | ||||
AK303384.1 | ||||
BC136636.1 | ||||
BC144328.1 | ||||
BC150190.1 | ||||
D01038.1 | BAA00845.1 | |||
DA655978.1 | ||||
M59911.1 | AAA36120.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509660 | Chr.17:50020085 - 50083399 on Build GRCh38 | Insertion |
![]() ![]() |
nsv833474 | Chr.17:49861663 - 50094199 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)