Assay Details
Target Gene Details
Entrez Gene ID: | 23135 |
Gene Name: | lysine demethylase 6B |
Gene Aliases: |
JMJD3 |
Location: |
Chr.17:7834210-7854800 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KDM6B | XM_005256549.3 | XP_005256606.1 | ||
XM_005256551.3 | XP_005256608.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833351 | Chr.17:7755691 - 7893863 on Build GRCh38 | Loss | NAA38 DNAH2 KDM6B CYB5D1 CHD3 TMEM88 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |