Assay Details
Target Gene Details
Entrez Gene ID: | 9487 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class L |
Gene Aliases: |
CHIME |
Location: |
Chr.17:16217195-16351800 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIGL | NM_004278.3 | NP_004269.1 | ||
XM_011524080.2 | XP_011522382.1 | |||
XM_017025349.1 | XP_016880838.1 | |||
XM_017025350.1 | XP_016880839.1 | |||
XM_017025351.1 | XP_016880840.1 | |||
XM_017025352.1 | XP_016880841.1 | |||
XM_017025353.1 | XP_016880842.1 | |||
XM_017025354.1 | XP_016880843.1 | |||
XM_017025355.1 | XP_016880844.1 | |||
XM_017025356.1 | XP_016880845.1 | |||
AB017165.1 | BAA74775.1 | |||
AK292932.1 | ||||
AK296120.1 | ||||
AK302523.1 | ||||
BC068197.1 | AAH68197.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv528263 | Chr.17:16194116 - 16293682 on Build GRCh38 | Loss | NCOR1 PIGL MIR1288 |
esv2715702 | Chr.17:15750514 - 16668268 on Build GRCh38 | Deletion | ZSWIM7 LRRC75A-AS1 NCOR1 PIGL TRPV2 ZNF624 CCDC144A CDRT15P2 MEIS3P1 ADORA2B SNORD65 CENPV ZNF287 SNORD49A TTC19 LRRC75A LOC101928567 MIR1288 SNORD49B UBB |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |