Assay Details
Target Gene Details
Entrez Gene ID: | 9358 |
Gene Name: | integrin subunit beta like 1 |
Gene Aliases: |
OSCP, TIED |
Location: |
Chr.13:101452593-101720857 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ITGBL1 | NM_001271754.1 | NP_001258683.1 | ||
NM_001271755.1 | NP_001258684.1 | |||
NM_004791.2 | NP_004782.1 | |||
XM_005254100.4 | XP_005254157.1 | |||
AB008375.1 | BAA22994.1 | |||
AF072752.1 | AAD17666.1 | |||
AK095875.1 | ||||
AK291340.1 | ||||
AK297744.1 | ||||
AK308194.1 | ||||
BC036788.1 | AAH36788.2 | |||
BP284376.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv579e212 | Chr.13:101470630 - 101481231 on Build GRCh38 | Loss |
![]() |
nsv832701 | Chr.13:101445422 - 101614006 on Build GRCh38 | Loss |
![]() |
esv3633363 | Chr.13:101469479 - 101481605 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)