Assay Details
Target Gene Details
Entrez Gene ID: | 100862704 |
Gene Name: | long intergenic non-protein coding RNA 441 |
Gene Aliases: |
ncRNA-RB1 |
Location: |
Chr.13:48296513-48303661 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC00441 | NR_046414.1 | 1 | 244 |