Assay Details
Target Gene Details
Entrez Gene ID: | 282706 |
Gene Name: | DAOA antisense RNA 1 |
Gene Aliases: |
DAOA-AS, DAOAAS, G30 |
Location: |
Chr.13:105459055-105505681 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DAOA-AS1 | NR_040247.1 | 7 | 2192 | |
AY138548.1 | 7 | 2192 | AAN08434.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv562996 | Chr.13:104426430 - 105664629 on Build GRCh38 | Gain | DAOA DAOA-AS1 |
esv3633445 | Chr.13:105457162 - 105460959 on Build GRCh38 | Loss | DAOA-AS1 |