Assay Details
Target Gene Details
Entrez Gene ID: | 55739 |
Gene Name: | NAD(P)HX dehydratase |
Gene Aliases: |
CARKD, LP3298 |
Location: |
Chr.13:110615460-110639996 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAXD | NM_001242881.1 | NP_001229810.1 | ||
NM_001242882.1 | NP_001229811.1 | |||
NM_001242883.1 | NP_001229812.1 | |||
NM_018210.3 | NP_060680.2 | |||
NR_040103.1 | ||||
NR_040104.1 | ||||
AK001631.1 | BAA91797.1 | |||
AK024260.1 | BAB14863.1 | |||
AK057767.1 | ||||
AK093771.1 | ||||
AK225129.1 | ||||
AK296227.1 | ||||
AK296758.1 | ||||
AK298917.1 | ||||
AK302117.1 | ||||
BC041028.1 | AAH41028.1 | |||
BQ955656.1 | ||||
DA175011.1 | ||||
DA435066.1 | ||||
DA815628.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1044234 | Chr.13:110490706 - 110718233 on Build GRCh38 | Gain | COL4A2-AS1 NAXD COL4A2 LOC105376684 CARS2 RAB20 ING1 |
nsv563124 | Chr.13:110608920 - 110648596 on Build GRCh38 | Loss | NAXD CARS2 |
nsv1046423 | Chr.13:110283757 - 110634260 on Build GRCh38 | Gain | MIR8073 COL4A2-AS1 NAXD COL4A1 COL4A2-AS2 COL4A2 RAB20 |
nsv563123 | Chr.13:110584913 - 110639697 on Build GRCh38 | Loss | NAXD |
nsv832720 | Chr.13:110447888 - 110643646 on Build GRCh38 | Loss | COL4A2-AS1 NAXD COL4A2-AS2 COL4A2 CARS2 RAB20 |
nsv1044340 | Chr.13:110581726 - 111040647 on Build GRCh38 | Gain | LOC105370363 LINC00346 ANKRD10 NAXD LOC105376684 LINC00431 CARS2 ING1 LOC105370362 LINC00567 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |