Assay Details
Target Gene Details
Entrez Gene ID: | 2700 |
Gene Name: | gap junction protein alpha 3 |
Gene Aliases: |
CTRCT14, CX46, CZP3 |
Location: |
Chr.13:20138252-20161327 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GJA3 | NM_021954.3 | NP_068773.2 | ||
XM_011535048.2 | XP_011533350.1 | |||
DR003755.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3631421 | Chr.13:20145912 - 20223722 on Build GRCh38 | Gain | GJB6 GJB2 GJA3 |
nsv1046946 | Chr.13:19730301 - 20507454 on Build GRCh38 | Gain | GJB6 PSPC1 GJB2 LINC01072 MIR4499 GJA3 ZMYM5 ZMYM2 CRYL1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |