Assay Details
Target Gene Details
Entrez Gene ID: | 100190939 |
Gene Name: | TPT1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:45341345-45391483 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TPT1-AS1 | NR_024458.1 | |||
AF318337.1 | AAL55844.1 | |||
AK124332.1 | BAC85835.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3631951 | Chr.13:45352364 - 45361056 on Build GRCh38 | Loss | TPT1-AS1 |
nsv510579 | Chr.13:45305226 - 45424938 on Build GRCh38 | Deletion | SLC25A30-AS1 SNORA31 TPT1-AS1 SLC25A30 TPT1 |
esv3580625 | Chr.13:45348477 - 45359056 on Build GRCh38 | Loss | TPT1-AS1 |