Assay Details
Target Gene Details
Entrez Gene ID: | 3356 |
Gene Name: | 5-hydroxytryptamine receptor 2A |
Gene Aliases: |
5-HT2A, HTR2 |
Location: |
Chr.13:46831542-46897076 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HTR2A | NM_000621.4 | NP_000612.1 | ||
NM_001165947.2 | NP_001159419.1 | |||
A23337.1 | CAA01675.1 | |||
AF498982.1 | AAM21129.1 | |||
AK302787.1 | ||||
AK314132.1 | ||||
BC069356.1 | AAH69356.1 | |||
BC069576.1 | AAH69576.1 | |||
BC074848.2 | AAH74848.1 | |||
BC074849.2 | AAH74849.1 | |||
BC096839.1 | AAH96839.1 | |||
BX647777.1 | ||||
M86841.1 | AAA58354.1 | |||
S71229.1 | AAB31320.1 | |||
X57830.1 | CAA40963.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2763019 | Chr.13:46774094 - 47549312 on Build GRCh38 | Loss | HTR2A HTR2A-AS1 ESD |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |