Assay Details
Target Gene Details
Entrez Gene ID: | 92293 |
Gene Name: | transmembrane protein 132C |
Gene Aliases: |
PPP1R152 |
Location: |
Chr.12:128267170-128707915 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM132C | NM_001136103.2 | NP_001129575.2 | ||
XM_011538998.2 | XP_011537300.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1573n100 | Chr.12:126502891 - 128427644 on Build GRCh38 | Gain | LINC00943 LINC00507 TMEM132C MIR3612 LOC440117 LOC101927616 FLJ37505 LINC00944 LOC101927592 LOC101927694 LOC100996679 LOC107984449 CRAT8 LINC00508 LOC101927637 LOC105370068 MIR4419B LOC107984465 LOC100996671 |
nsv1160070 | Chr.12:128223477 - 128299998 on Build GRCh38 | Duplication | TMEM132C MIR3612 MIR4419B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |