Assay Details
Target Gene Details
Entrez Gene ID: | 121506 |
Gene Name: | endoplasmic reticulum protein 27 |
Gene Aliases: |
C12orf46, PDIA8 |
Location: |
Chr.12:14914027-14938549 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ERP27 | NM_152321.3 | NP_689534.1 | ||
AK056677.1 | BAB71251.1 | |||
AY358536.1 | AAQ88900.1 | |||
BC030218.1 | AAH30218.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv469120 | Chr.12:14922268 - 14948501 on Build GRCh38 | Gain | ARHGDIB ERP27 |
nsv557612 | Chr.12:14921379 - 14939445 on Build GRCh38 | Gain | ERP27 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |