Assay Details
Target Gene Details
Entrez Gene ID: | 171017 |
Gene Name: | zinc finger protein 384 |
Gene Aliases: |
CAGH1, CAGH1A, CIZ, ERDA2, NMP4, NP, TNRC1 |
Location: |
Chr.12:6666477-6689572 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv983480 | Chr.12:6670944 - 6674895 on Build GRCh38 | Duplication | ZNF384 |
nsv832322 | Chr.12:6575884 - 6765914 on Build GRCh38 | Loss | CHD4 ACRBP ING4 MLF2 LPAR5 COPS7A SCARNA11 ZNF384 PIANP |
nsv832323 | Chr.12:6662233 - 6856559 on Build GRCh38 | Loss | CD4 GPR162 MLF2 USP5 PTMS ZNF384 CDCA3 PIANP LAG3 P3H3 ING4 GNB3 COPS7A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |