Assay Details
Target Gene Details
Entrez Gene ID: | 5939 |
Gene Name: | RNA binding motif single stranded interacting protein 2 |
Gene Aliases: |
SCR3 |
Location: |
Chr.12:56521812-56596196 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RBMS2 | NM_002898.3 | NP_002889.1 | ||
XM_005269059.4 | XP_005269116.1 | |||
XM_005269060.4 | XP_005269117.1 | |||
XM_005269061.3 | XP_005269118.1 | |||
XM_005269066.4 | XP_005269123.1 | |||
XM_006719541.3 | XP_006719604.1 | |||
XM_006719542.3 | XP_006719605.1 | |||
XM_006719543.3 | XP_006719606.1 | |||
XM_006719544.3 | XP_006719607.1 | |||
XM_011538637.2 | XP_011536939.1 | |||
XM_011538638.2 | XP_011536940.1 | |||
XM_011538639.2 | XP_011536941.1 | |||
XM_011538640.2 | XP_011536942.1 | |||
XM_017019775.1 | XP_016875264.1 | |||
AK293315.1 | ||||
AK299580.1 | ||||
AK304540.1 | ||||
AK308885.1 | ||||
BC027863.1 | AAH27863.1 | |||
BC072679.1 | AAH72679.1 | |||
D28483.1 | BAA05842.1 | |||
DC382164.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss | ZBTB39 PRIM1 MIR1228 COQ10A PTGES3 RBMS2 R3HDM2 CS GLS2 NXPH4 TIMELESS DDIT3 DTX3 SLC39A5 NEMP1 DCTN2 SHMT2 KIF5A MIR616 SNORD59B PIP4K2C ARHGAP9 LRP1 HSD17B6 ARHGEF25 NDUFA4L2 ATP5B RNF41 NACA NABP2 INHBE STAT6 GLI1 MARS MIP STAT2 HBCBP APOF SPRYD4 TAC3 ANKRD52 RDH16 SDR9C7 OS9 PAN2 STAC3 B4GALNT1 SLC26A10 MBD6 GPR182 INHBC MIR6758 MYO1A CNPY2 BAZ2A LRP1-AS SNORA105C SNORD59A NAB2 LOC101927583 IL23A |
nsv1143564 | Chr.12:56544116 - 56549916 on Build GRCh38 | Deletion | RBMS2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |