Assay Details
Target Gene Details
Entrez Gene ID: | 9478 |
Gene Name: | calcium binding protein 1 |
Gene Aliases: |
CALBRAIN, HCALB_BR |
Location: |
Chr.12:120640552-120680869 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CABP1 | NM_001033677.1 | NP_001028849.1 | ||
NM_004276.4 | NP_004267.2 | |||
NM_031205.3 | NP_112482.1 | |||
XM_017020235.1 | XP_016875724.1 | |||
XM_017020238.1 | XP_016875727.1 | |||
XM_017020239.1 | XP_016875728.1 | |||
AF169148.1 | AAF25782.1 | |||
AF169149.1 | AAF25783.1 | |||
AL533429.3 | ||||
BC030201.1 | ||||
BM716422.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1045923 | Chr.12:120644400 - 120708416 on Build GRCh38 | Gain | MLEC CABP1 |
nsv521928 | Chr.12:120603925 - 121356945 on Build GRCh38 | Gain | SPPL3 CAMKK2 P2RX7 HNF1A MLEC UNC119B CABP1 P2RX4 ACADS HNF1A-AS1 MIR4700 ANAPC5 OASL XLOC_009911 C12orf43 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |