Assay Details
Target Gene Details
Entrez Gene ID: | 5025 |
Gene Name: | purinergic receptor P2X 4 |
Gene Aliases: |
P2X4, P2X4R |
Location: |
Chr.12:121209861-121234106 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
P2RX4 | NM_001256796.1 | NP_001243725.1 | ||
NM_001261397.1 | NP_001248326.1 | |||
NM_001261398.1 | NP_001248327.1 | |||
NM_002560.2 | NP_002551.2 | |||
NR_046372.1 | ||||
NR_046373.1 | ||||
XM_011538416.2 | XP_011536718.1 | |||
AF000234.1 | AAB58405.1 | |||
AF012903.1 | AAB66834.1 | |||
AK225604.1 | ||||
AK293599.1 | ||||
AK293660.1 | ||||
AK297704.1 | ||||
AK301080.1 | ||||
AK308549.1 | ||||
AK308868.1 | ||||
BC033826.1 | AAH33826.1 | |||
BT019738.1 | AAV38543.1 | |||
BT019739.1 | AAV38544.1 | |||
BX402887.2 | ||||
U83993.1 | AAD00553.1 | |||
Y07684.1 | CAA68948.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv560455 | Chr.12:121087788 - 121363295 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv1054878 | Chr.12:121082077 - 121328424 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv560454 | Chr.12:121019270 - 121307939 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv521928 | Chr.12:120603925 - 121356945 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)