Assay Details
Target Gene Details
Entrez Gene ID: | 9182 |
Gene Name: | Ras association domain family member 9 |
Gene Aliases: |
P-CIP1, PAMCI, PCIP1 |
Location: |
Chr.12:85800703-85836540 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RASSF9 | NM_005447.3 | NP_005438.2 | ||
XM_011538988.2 | XP_011537290.1 | |||
AF056209.1 | AAD03250.1 | |||
AK022089.1 | ||||
BC031589.1 | AAH31589.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1539n100 | Chr.12:85792109 - 85878715 on Build GRCh38 | Gain | RASSF9 NTS |
nsv1049179 | Chr.12:85662598 - 86405410 on Build GRCh38 | Gain | RASSF9 NTS MGAT4C |
esv3892242 | Chr.12:85514618 - 85885975 on Build GRCh38 | Loss | RASSF9 NTS |
dgv1537n100 | Chr.12:85264824 - 85841655 on Build GRCh38 | Loss | RASSF9 LRRIQ1 ALX1 |
esv3892241 | Chr.12:85305039 - 88102633 on Build GRCh38 | Gain | LOC105369879 RASSF9 CEP290 NTS MGAT4C MKRN9P C12orf29 C12orf50 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |