Assay Details
Target Gene Details
Entrez Gene ID: | 5939 |
Gene Name: | RNA binding motif single stranded interacting protein 2 |
Gene Aliases: |
SCR3 |
Location: |
Chr.12:56521812-56596196 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RBMS2 | NM_002898.3 | NP_002889.1 | ||
XM_005269059.4 | XP_005269116.1 | |||
XM_005269060.4 | XP_005269117.1 | |||
XM_005269061.3 | XP_005269118.1 | |||
XM_005269066.4 | XP_005269123.1 | |||
XM_006719541.3 | XP_006719604.1 | |||
XM_006719542.3 | XP_006719605.1 | |||
XM_006719543.3 | XP_006719606.1 | |||
XM_006719544.3 | XP_006719607.1 | |||
XM_011538638.2 | XP_011536940.1 | |||
XM_011538639.2 | XP_011536941.1 | |||
XM_011538640.2 | XP_011536942.1 | |||
AK293315.1 | ||||
AK299580.1 | ||||
AK304540.1 | ||||
AK308885.1 | ||||
BC027863.1 | AAH27863.1 | |||
BC072679.1 | AAH72679.1 | |||
D28483.1 | BAA05842.1 | |||
DC382164.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss | MIR6758 SLC39A5 IL23A GLI1 R3HDM2 PAN2 LOC101927583 MBD6 LRP1-AS SLC26A10 GPR182 APOF SDR9C7 RBMS2 ARHGEF25 ANKRD52 KIF5A TAC3 MYO1A STAT6 RDH16 CNPY2 ZBTB39 HBCBP STAC3 ARHGAP9 SNORA105C STAT2 CS MIR1228 TIMELESS LRP1 SPRYD4 ATP5B NEMP1 B4GALNT1 SNORD59A NAB2 INHBE DCTN2 NABP2 MIP GLS2 MARS NXPH4 HSD17B6 DTX3 SHMT2 PRIM1 SNORD59B PIP4K2C COQ10A INHBC PTGES3 NDUFA4L2 OS9 MIR616 NACA RNF41 DDIT3 BAZ2A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |