Assay Details
Target Gene Details
Entrez Gene ID: | 10818 |
Gene Name: | fibroblast growth factor receptor substrate 2 |
Gene Aliases: |
FRS1A, FRS2A, FRS2alpha, SNT, SNT-1, SNT1 |
Location: |
Chr.12:69470349-69579793 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FRS2 | NM_001042555.2 | NP_001036020.1 | ||
NM_001278351.1 | NP_001265280.1 | |||
NM_001278353.1 | NP_001265282.1 | |||
NM_001278354.1 | NP_001265283.1 | |||
NM_001278355.1 | NP_001265284.1 | |||
NM_001278356.1 | NP_001265285.1 | |||
NM_001278357.1 | NP_001265286.1 | |||
NM_006654.4 | NP_006645.3 | |||
XM_017018717.1 | XP_016874206.1 | |||
XM_017018718.1 | XP_016874207.1 | |||
AK312477.1 | ||||
BC021562.1 | AAH21562.1 | |||
BP217337.1 | ||||
BP230333.1 | ||||
BP280775.1 | ||||
BP311537.1 | ||||
BP363404.1 | ||||
BU166503.1 | ||||
DA189114.1 | ||||
DA311708.1 | ||||
DA721439.1 | ||||
DB081843.1 | ||||
DR003634.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |