Assay Details
Target Gene Details
Entrez Gene ID: | 8496 |
Gene Name: | PPFIA binding protein 1 |
Gene Aliases: |
L2, SGT2, hSGT2, hSgt2p |
Location: |
Chr.12:27524112-27695564 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1406n100 | Chr.12:27128236 - 27640822 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
esv3892167 | Chr.12:27548131 - 27634686 on Build GRCh38 | Gain |
![]() |
dgv2417n54 | Chr.12:27068022 - 27638988 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv832358 | Chr.12:27519016 - 27705439 on Build GRCh38 | Gain |
![]() ![]() |
nsv436825 | Chr.12:27135971 - 27642579 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() |
esv2745664 | Chr.12:27017516 - 27820117 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3628940 | Chr.12:27451350 - 27631744 on Build GRCh38 | Gain |
![]() ![]() |
nsv521872 | Chr.12:27114021 - 27635541 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
dgv271e199 | Chr.12:27089700 - 27631451 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)