Assay Details
Target Gene Details
Entrez Gene ID: | 26127 |
Gene Name: | FGFR1 oncogene partner 2 |
Gene Aliases: |
HSPC123-like, WIT3.0 |
Location: |
Chr.12:26938372-26966648 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FGFR1OP2 | NM_001171887.1 | NP_001165358.1 | ||
NM_001171888.1 | NP_001165359.1 | |||
NM_015633.2 | NP_056448.1 | |||
AF161472.1 | AAF29087.1 | |||
AK001534.1 | BAA91745.1 | |||
BC032143.2 | AAH32143.1 | |||
BI829470.1 | ||||
DA388639.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv557804 | Chr.12:26945269 - 26966684 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)