Assay Details
Target Gene Details
Entrez Gene ID: | 5331 |
Gene Name: | phospholipase C beta 3 |
Gene Aliases: |
- |
Location: |
Chr.11:64251523-64269452 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLCB3 | NM_000932.2 | NP_000923.1 | ||
NM_001184883.1 | NP_001171812.1 | |||
NM_001316314.1 | NP_001303243.1 | |||
XM_011545101.2 | XP_011543403.1 | |||
XM_017017925.1 | XP_016873414.1 | |||
BC032659.1 | AAH32659.1 | |||
BC142681.1 | ||||
CN278806.1 | ||||
Z26649.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1159904 | Chr.11:64150129 - 64279169 on Build GRCh38 | Deletion | DNAJC4 FKBP2 BAD FERMT3 NUDT22 LOC105369340 TRPT1 PPP1R14B PLCB3 MACROD1 VEGFB GPR137 STIP1 |
nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion | PRDX5 FERMT3 ESRRA KCNK4-TEX40 TRPT1 NAA40 PPP1R14B PLCB3 TRMT112 MIR7155 TEX40 STIP1 DNAJC4 FKBP2 OTUB1 BAD NUDT22 LOC105369340 KCNK4 CCDC88B FLRT1 COX8A MACROD1 VEGFB GPR137 |
dgv1963n54 | Chr.11:64174262 - 64329761 on Build GRCh38 | Loss | DNAJC4 FKBP2 BAD PRDX5 FERMT3 ESRRA NUDT22 LOC105369340 KCNK4-TEX40 TRPT1 KCNK4 PPP1R14B PLCB3 TRMT112 VEGFB GPR137 TEX40 STIP1 |
nsv555187 | Chr.11:64185649 - 64260416 on Build GRCh38 | Loss | DNAJC4 FKBP2 PLCB3 FERMT3 NUDT22 LOC105369340 TRPT1 VEGFB STIP1 PPP1R14B |
nsv832186 | Chr.11:64204788 - 64402587 on Build GRCh38 | Loss | DNAJC4 FKBP2 BAD PRDX5 FERMT3 ESRRA NUDT22 LOC105369340 KCNK4-TEX40 TRPT1 KCNK4 CCDC88B PPP1R14B PLCB3 MIR1237 TRMT112 VEGFB MIR7155 GPR137 TEX40 RPS6KA4 |
nsv555184 | Chr.11:64156974 - 64275525 on Build GRCh38 | Loss | DNAJC4 FKBP2 BAD FERMT3 NUDT22 LOC105369340 TRPT1 PPP1R14B PLCB3 MACROD1 VEGFB GPR137 STIP1 |
nsv469961 | Chr.11:64055468 - 64318495 on Build GRCh38 | Loss | DNAJC4 FKBP2 BAD PRDX5 FERMT3 ESRRA NUDT22 LOC105369340 KCNK4-TEX40 TRPT1 KCNK4 PPP1R14B FLRT1 PLCB3 MACROD1 TRMT112 VEGFB GPR137 TEX40 STIP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |