Assay Details
Target Gene Details
Entrez Gene ID: | 114609 |
Gene Name: | TIR domain containing adaptor protein |
Gene Aliases: |
BACTS1, Mal, MyD88-2, wyatt |
Location: |
Chr.11:126281268-126294933 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TIRAP | NM_001039661.1 | NP_001034750.1 | ||
NM_001318776.1 | NP_001305705.1 | |||
NM_001318777.1 | NP_001305706.1 | |||
NM_148910.2 | NP_683708.1 | |||
XM_005271399.3 | XP_005271456.2 | |||
XM_011542576.2 | XP_011540878.1 | |||
XM_017017161.1 | XP_016872650.1 | |||
XM_017017162.1 | XP_016872651.1 | |||
AF378129.1 | AAL05627.1 | |||
AF410783.1 | AAL05036.1 | |||
AK124298.1 | ||||
AK313147.1 | ||||
BC032474.1 | AAH32474.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv556530 | Chr.11:126264724 - 126393990 on Build GRCh38 | Loss | ST3GAL4-AS1 HP11014 FOXRED1 ST3GAL4 DCPS SRPRA TIRAP |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |