Assay Details
Target Gene Details
Entrez Gene ID: | 387804 |
Gene Name: | V-set and transmembrane domain containing 5 |
Gene Aliases: |
C11orf90 |
Location: |
Chr.11:93817315-93850653 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VSTM5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3627294 | Chr.11:93485306 - 93944317 on Build GRCh38 | Gain | SNORD5 SNORA32 CEP295 SNORA25 SNORA18 SNORA8 SNORA40 SNORD6 SMCO4 C11orf54 MED17 VSTM5 MIR1304 SNORA1 TAF1D SCARNA9 |