Assay Details
Target Gene Details
Entrez Gene ID: | 60529 |
Gene Name: | ALX homeobox 4 |
Gene Aliases: |
CRS5, FND2 |
Location: |
Chr.11:44260728-44310166 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALX4 | NM_021926.3 | NP_068745.2 | ||
AB058691.1 | ||||
AF294629.1 | AAG23961.1 | |||
AJ404888.1 | CAC15060.1 | |||
AK092346.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832139 | Chr.11:44063681 - 44277647 on Build GRCh38 | Loss | ALX4 ACCS EXT2 |
dgv67n21 | Chr.11:44250972 - 44277071 on Build GRCh38 | Loss | ALX4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |