Assay Details
Target Gene Details
Entrez Gene ID: | 84866 |
Gene Name: | transmembrane protein 25 |
Gene Aliases: |
- |
Location: |
Chr.11:118531088-118546670 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM25 | NM_001144037.1 | NP_001137509.1 | ||
NM_001144038.1 | NP_001137510.1 | |||
XM_017018431.1 | XP_016873920.1 | |||
AK027305.1 | BAB55029.1 | |||
AK075437.1 | BAC11620.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1044281 | Chr.11:118503814 - 118554704 on Build GRCh38 | Gain | IFT46 TMEM25 TTC36 KMT2A LOC101929089 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |