Assay Details
Target Gene Details
Entrez Gene ID: | 79829 |
Gene Name: | N(alpha)-acetyltransferase 40, NatD catalytic subunit |
Gene Aliases: |
NAT11, PATT1 |
Location: |
Chr.11:63938970-63957322 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAA40 | NM_001300800.1 | NP_001287729.1 | ||
NM_024771.3 | NP_079047.2 | |||
XM_006718689.3 | XP_006718752.1 | |||
XM_011545254.2 | XP_011543556.1 | |||
XM_011545255.2 | XP_011543557.1 | |||
AK023910.1 | BAB14720.1 | |||
AK299041.1 | ||||
AK300451.1 | ||||
AK303557.1 | ||||
BC041617.1 | AAH41617.1 | |||
BC052298.1 | AAH52298.1 | |||
BX647309.1 | CAI46006.1 | |||
DA649381.1 | ||||
DC307984.1 | ||||
HY009487.1 | ||||
JF263566.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv555180 | Chr.11:63748280 - 63945767 on Build GRCh38 | Loss | MARK2 RCOR2 C11orf84 NAA40 C11orf95 RTN3 |
nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion | TRPT1 FLRT1 COX8A OTUB1 FKBP2 LOC105369340 BAD PPP1R14B TEX40 GPR137 TRMT112 KCNK4-TEX40 NUDT22 PLCB3 STIP1 MACROD1 CCDC88B KCNK4 VEGFB NAA40 DNAJC4 ESRRA PRDX5 MIR7155 FERMT3 |
nsv555181 | Chr.11:63872409 - 64015880 on Build GRCh38 | Loss | MARK2 RCOR2 NAA40 COX8A OTUB1 MACROD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |