Assay Details
Target Gene Details
Entrez Gene ID: | 28971 |
Gene Name: | adipogenesis associated Mth938 domain containing |
Gene Aliases: |
C11orf67, CK067, PTD015 |
Location: |
Chr.11:77821114-77901281 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AAMDC | NM_001316957.1 | NP_001303886.1 | ||
NM_001316958.1 | NP_001303887.1 | |||
NM_001316960.1 | NP_001303889.1 | |||
NM_001316961.1 | NP_001303890.1 | |||
NM_001316962.1 | NP_001303891.1 | |||
NM_024684.3 | NP_078960.1 | |||
NR_133637.1 | ||||
XM_011544968.2 | XP_011543270.1 | |||
XM_011544969.2 | XP_011543271.1 | |||
XM_017017620.1 | XP_016873109.1 | |||
AF092136.1 | AAD40378.1 | |||
AI569731.1 | ||||
AK024688.1 | BAB14963.1 | |||
BC002752.2 | AAH02752.1 | |||
BC016854.2 | AAH16854.1 | |||
BI791786.1 | ||||
BM930331.1 | ||||
BP270361.1 | ||||
BX646013.1 | ||||
CN362204.1 | ||||
CR457332.1 | CAG33613.1 |
Target Gene Details
Entrez Gene ID: | 51773 |
Gene Name: | remodeling and spacing factor 1 |
Gene Aliases: |
HBXAP, RSF-1, XAP8, p325 |
Location: |
Chr.11:77660009-77872232 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RSF1 | XM_017017923.1 | XP_016873412.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv520740 | Chr.11:77842592 - 78286890 on Build GRCh38 | Gain | KCTD21-AS1 INTS4 KCTD21 NDUFC2-KCTD14 USP35 RSF1 GAB2 NDUFC2 THRSP ALG8 KCTD14 AAMDC |
nsv1050891 | Chr.11:77705600 - 78076956 on Build GRCh38 | Gain | INTS4 NDUFC2-KCTD14 RSF1 NDUFC2 THRSP KCTD14 RPS20P27 AAMDC |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |