Assay Details
Target Gene Details
Entrez Gene ID: | 272 |
Gene Name: | adenosine monophosphate deaminase 3 |
Gene Aliases: |
- |
Location: |
Chr.11:10450321-10507579 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AMPD3 | NM_000480.2 | NP_000471.1 | ||
NM_001025389.1 | NP_001020560.1 | |||
NM_001025390.1 | NP_001020561.1 | |||
NM_001172431.1 | NP_001165902.1 | |||
AK289998.1 | ||||
AK294444.1 | ||||
AK295046.1 | ||||
AK299113.1 | ||||
AK301507.1 | ||||
AK308181.1 | ||||
AK311436.1 | ||||
AK311711.1 | ||||
AK313039.1 | ||||
BC126118.1 | ||||
D12775.1 | BAA02240.1 | |||
DC334250.1 | ||||
M84720.1 | AAA58365.1 | |||
M84721.1 | AAA58366.1 | |||
M84722.1 | AAA58367.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3625358 | Chr.11:10435119 - 10512909 on Build GRCh38 | Gain | AMPD3 MIR4485 MTRNR2L8 RNF141 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |