Assay Details
Target Gene Details
Entrez Gene ID: | 63876 |
Gene Name: | PBX/knotted 1 homeobox 2 |
Gene Aliases: |
PREP2 |
Location: |
Chr.11:125111997-125433389 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PKNOX2 | XM_017018110.1 | XP_016873599.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv556488 | Chr.11:125111809 - 125296916 on Build GRCh38 | Gain | PKNOX2 PKNOX2-AS1 KRT18P59 |
nsv832290 | Chr.11:125112706 - 125271154 on Build GRCh38 | Gain+Loss | PKNOX2 PKNOX2-AS1 KRT18P59 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |