Assay Details
Target Gene Details
Entrez Gene ID: | 50862 |
Gene Name: | ring finger protein 141 |
Gene Aliases: |
ZFP26, ZNF230 |
Location: |
Chr.11:10511678-10541227 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RNF141 | NM_016422.3 | NP_057506.2 | ||
AF214680.1 | AAF30180.1 | |||
AK289764.1 | ||||
AK309367.1 | ||||
AL598903.1 | ||||
BC018104.1 | AAH18104.1 | |||
BT006662.1 | AAP35308.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1044011 | Chr.11:10507806 - 10542903 on Build GRCh38 | Gain | MIR4485 RNF141 MRVI1-AS1 MTRNR2L8 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |