Assay Details
Target Gene Details
Entrez Gene ID: | 11309 |
Gene Name: | solute carrier organic anion transporter family member 2B1 |
Gene Aliases: |
OATP-B, OATP2B1, OATPB, SLC21A9 |
Location: |
Chr.11:75150987-75206400 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLCO2B1 | NM_001145212.2 | NP_001138684.1 | ||
NM_007256.4 | NP_009187.1 | |||
AB020687.1 | ||||
AB026256.1 | BAA78638.1 | |||
AK290234.1 | ||||
AK300134.1 | ||||
AK302492.1 | ||||
AK304783.1 | ||||
AL117465.1 | CAB55940.1 | |||
BC041095.1 | AAH41095.1 | |||
DC335261.1 | ||||
DC343392.1 | ||||
FM209049.1 | ||||
FM209050.1 | ||||
FM209051.1 | ||||
FM209052.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |