Assay Details
Target Gene Details
Entrez Gene ID: | 1643 |
Gene Name: | damage specific DNA binding protein 2 |
Gene Aliases: |
DDBB, UV-DDB2, XPE |
Location: |
Chr.11:47214942-47239218 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDB2 | NM_000107.2 | NP_000098.1 | ||
NM_001300734.1 | NP_001287663.1 | |||
AB107037.1 | BAD12557.1 | |||
AB107038.1 | BAD12558.1 | |||
AB107039.1 | BAD12559.1 | |||
AB107040.1 | BAD12560.1 | |||
AK091640.1 | ||||
AK313262.1 | ||||
BC000093.2 | AAH00093.1 | |||
BT007139.1 | AAP35803.1 | |||
U18300.1 | AAB07897.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_next.png)
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_next.png)
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_next.png)
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)