Assay Details
Target Gene Details
Entrez Gene ID: | 89797 |
Gene Name: | neuron navigator 2 |
Gene Aliases: |
HELAD1, POMFIL2, RAINB1, STEERIN2, UNC53H2 |
Location: |
Chr.11:19345200-20121601 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAV2 | NM_001111018.1 | NP_001104488.1 | ||
XM_011520452.2 | XP_011518754.1 | |||
XM_017018520.1 | XP_016874009.1 | |||
XM_017018522.1 | XP_016874011.1 | |||
AB063115.1 | BAC00853.1 |
Target Gene Details
Entrez Gene ID: | 399876 |
Gene Name: | NAV2 antisense RNA 4 |
Gene Aliases: |
- |
Location: |
Chr.11:19510893-19524078 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAV2-AS4 | NR_033989.1 | |||
AK023501.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3891939 | Chr.11:19507092 - 19565555 on Build GRCh38 | Gain | NAV2-AS4 NAV2-AS5 NAV2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |