Assay Details
Target Gene Details
Entrez Gene ID: | 7275 |
Gene Name: | tubby bipartite transcription factor |
Gene Aliases: |
RDOB, rd5 |
Location: |
Chr.11:8019180-8106107 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TUB | NM_003320.4 | NP_003311.2 | ||
XM_005253109.3 | XP_005253166.1 | |||
AK293528.1 | ||||
BC075031.2 | AAH75031.1 | |||
BC075032.2 | AAH75032.1 | |||
U82467.1 | AAB53699.1 |
Target Gene Details
Entrez Gene ID: | 101927917 |
Gene Name: | TUB antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:8060038-8069373 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TUB-AS1 | NR_120537.1 | |||
NR_120538.1 | ||||
AK055772.1 | ||||
DA626152.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428249 | Chr.11:8047047 - 8194822 on Build GRCh38 | Loss | LOC101927936 TUB-AS1 TUB RIC3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |