Assay Details
Target Gene Details
Entrez Gene ID: | 3145 |
Gene Name: | hydroxymethylbilane synthase |
Gene Aliases: |
PBG-D, PBGD, PORC, UPS |
Location: |
Chr.11:119084871-119093549 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HMBS | NM_000190.3 | NP_000181.2 | ||
NM_001258208.1 | NP_001245137.1 | |||
NM_001258209.1 | NP_001245138.1 | |||
XM_005271531.1 | XP_005271588.1 | |||
XM_005271532.1 | XP_005271589.1 | |||
XM_005271533.3 | XP_005271590.1 | |||
AK000628.1 | ||||
AK131072.1 | ||||
AK290275.1 | ||||
AK309544.1 | ||||
BC000520.2 | AAH00520.1 | |||
BC008149.1 | AAH08149.1 | |||
BC019323.1 | AAH19323.1 | |||
CB128006.1 | ||||
X04808.1 | CAA28499.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |