Assay Details
Target Gene Details
Entrez Gene ID: | 28971 |
Gene Name: | adipogenesis associated Mth938 domain containing |
Gene Aliases: |
C11orf67, CK067, PTD015 |
Location: |
Chr.11:77821114-77901281 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AAMDC | NM_001316957.1 | NP_001303886.1 | ||
NM_001316958.1 | NP_001303887.1 | |||
NR_133637.1 | ||||
XM_017017620.1 | XP_016873109.1 | |||
AF092136.1 | AAD40378.1 | |||
BC016854.2 | AAH16854.1 | |||
BP270361.1 | ||||
BX646013.1 | ||||
CN362204.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv520740 | Chr.11:77842592 - 78286890 on Build GRCh38 | Gain | KCTD21-AS1 NDUFC2-KCTD14 AAMDC GAB2 INTS4 KCTD21 USP35 RSF1 THRSP NDUFC2 ALG8 KCTD14 |
nsv524537 | Chr.11:77872220 - 77882175 on Build GRCh38 | Loss | AAMDC INTS4 RSF1 |
nsv555453 | Chr.11:77866448 - 77907395 on Build GRCh38 | Loss | AAMDC INTS4 RSF1 |
nsv1050891 | Chr.11:77705600 - 78076956 on Build GRCh38 | Gain | RPS20P27 NDUFC2-KCTD14 AAMDC INTS4 RSF1 THRSP NDUFC2 KCTD14 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |