Assay Details
Target Gene Details
Entrez Gene ID: | 372 |
Gene Name: | archain 1 |
Gene Aliases: |
COPD |
Location: |
Chr.11:118572363-118603033 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARCN1 | NM_001142281.1 | NP_001135753.1 | ||
NM_001655.4 | NP_001646.2 | |||
XM_005271542.3 | XP_005271599.1 | |||
AK304019.1 | ||||
AK312298.1 | ||||
BC064936.1 | AAH64936.1 | |||
BC093636.1 | AAH93636.1 | |||
BC093638.1 | AAH93638.1 | |||
BI560739.1 | ||||
BX640860.1 | CAE45922.1 | |||
DB216489.1 | ||||
X81197.1 | CAA57071.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832283 | Chr.11:118566790 - 118786484 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)