Assay Details
Target Gene Details
Entrez Gene ID: | 8836 |
Gene Name: | gamma-glutamyl hydrolase |
Gene Aliases: |
GH |
Location: |
Chr.8:63015079-63039051 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GGH | NM_003878.2 | NP_003869.1 | ||
AK290450.1 | ||||
BC025025.1 | AAH25025.1 | |||
U55206.1 | AAC05579.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1018879 | Chr.8:62369165 - 63025073 on Build GRCh38 | Gain | NKAIN3 GGH UG0898H09 |
nsv6227 | Chr.8:62979528 - 63024393 on Build GRCh38 | Deletion | NKAIN3 GGH UG0898H09 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |