Assay Details
Target Gene Details
Entrez Gene ID: | 9530 |
Gene Name: | BCL2 associated athanogene 4 |
Gene Aliases: |
BAG-4, SODD |
Location: |
Chr.8:38176588-38213301 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BAG4 | NM_001204878.1 | NP_001191807.1 | ||
NM_004874.3 | NP_004865.1 | |||
AF095194.2 | AAD16123.2 | |||
AF111116.1 | AAD05226.1 | |||
AK304072.1 | ||||
AK314020.1 | ||||
BC038505.1 | AAH38505.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv7162n100 | Chr.8:38159377 - 38249049 on Build GRCh38 | Gain | DDHD2 LSM1 BAG4 |
dgv7165n100 | Chr.8:38200929 - 38249049 on Build GRCh38 | Loss | DDHD2 BAG4 |
dgv7163n100 | Chr.8:38194164 - 38252697 on Build GRCh38 | Gain | DDHD2 BAG4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |