Assay Details
Target Gene Details
Entrez Gene ID: | 2796 |
Gene Name: | gonadotropin releasing hormone 1 |
Gene Aliases: |
GNRH, GRH, LHRH, LNRH |
Location: |
Chr.8:25419258-25425040 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GNRH1 | NM_000825.3 | NP_000816.4 | ||
NM_001083111.1 | NP_001076580.1 | |||
BC126437.1 | ||||
BC126463.1 | ||||
CB243067.1 | ||||
M12578.1 | AAA35916.1 | |||
X01059.1 | CAA25526.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759606 | Chr.8:25411509 - 25564618 on Build GRCh38 | Loss | DOCK5 GNRH1 CDCA2 KCTD9 |
dgv7148n100 | Chr.8:25410448 - 25473361 on Build GRCh38 | Gain | DOCK5 GNRH1 CDCA2 KCTD9 |
nsv6125 | Chr.8:25410366 - 25444678 on Build GRCh38 | Insertion | DOCK5 GNRH1 KCTD9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |