Assay Details
Target Gene Details
Entrez Gene ID: | 148 |
Gene Name: | adrenoceptor alpha 1A |
Gene Aliases: |
ADRA1C, ADRA1L1, ALPHA1AAR |
Location: |
Chr.8:26738113-26870994 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADRA1A | NM_001322503.1 | NP_001309432.1 | ||
NM_033303.4 | NP_150646.3 | |||
NR_136343.1 | ||||
XM_006716292.3 | XP_006716355.1 | |||
XM_017013094.1 | XP_016868583.1 | |||
XM_017013095.1 | XP_016868584.1 | |||
AY491775.1 | AAR84644.1 | |||
AY491776.1 | AAR84645.1 | |||
D32202.1 | BAA06901.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)